Order Entry
Germany
Orders LinkContactUsLinkComponent
6-Hydroxyindole ≥98%

6-Hydroxyindole ≥98%

Glass bottle for solids, 1 g
Catalog # H55951.03
Corrosive
Irritant
Environment
6-Hydroxyindole ≥98%
Catalog # H55951.03
CAS Number:  2380-86-1
6-Hydroxyindole ≥98%
6-Hydroxyindole ≥98%

Glass bottle for solids, 1 g

CAS Number:  2380-86-1
Price
153,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
  • CAS Number:2380-86-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152101
  • Molecular Weight:133.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Ambient
  • EINECS:417-020-4
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.