LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 50MM/330-450MM, LRC50X330-450V01 1 * 1 items
Catalog # 554-2391
Supplier: Cytiva (Formerly Pall Lab)