ContactUsLinkComponent
LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 10MM/80-200MM, LRC10X080-200V01 1 * 1 items
Catalog # 554-2364
undefined
LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 10MM/80-200MM, LRC10X080-200V01 1 * 1 items
Catalog # 554-2364