ContactUsLinkComponent
Ready-to-use 1 mL and 5 mL pre-packed PRC columns.Q and S HyperCel sorbents are composed of a rigid cellulose matrix with excellent flow properties and low backpressure, providing high dynamic binding capacity at short residence time. PRC COLUMN 5X50 1 * 1 items
Catalog # 518-0082
Ready-to-use 1 mL and 5 mL pre-packed PRC columns.Q and S HyperCel sorbents are composed of a rigid cellulose matrix with excellent flow properties and low backpressure, providing high dynamic binding capacity at short residence time. PRC COLUMN 5X50 1 * 1 items
Catalog # 518-0082
Frequently Bought Together