Orders LinkContactUsLinkComponent
Watchmakers' magnifiers
Watchmakers' magnifiers
Magnifier with aspheric lens, 25 mm
Catalog # ESCH1124110
Supplier:   ESCHENBACH OPTIK
Watchmakers' magnifiers
Catalog # ESCH1124110
Supplier:  ESCHENBACH OPTIK
Watchmakers' magnifiers
Watchmakers' magnifiers
Magnifier with aspheric lens, 25 mm
Price
32,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite
Specifications
  • Ø:
    25 mm
  • Magnification:
    10×
  • Description:
    Magnifier with aspheric lens
  • Seq:
    003