Order Entry
Germany
Orders LinkContactUsLinkComponent
Spore strips

Spore strips

Bacillus atrophaeus cell line 9372: traditional spore strip 106 (24 month shelf life) / sterilisation process: dry heat, 30 mm, 6 mm
Catalog # NAMSBG-106DH
Supplier:   CROSSTEX
Spore strips
Catalog # NAMSBG-106DH
Supplier:  CROSSTEX
Spore strips

Bacillus atrophaeus cell line 9372: traditional spore strip 106 (24 month shelf life) / sterilisation process: dry heat, 30 mm, 6 mm

Price
337,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Documents

Certificates
Search by Lot Number
Specifications
  • Width:
    6 mm
  • Description:
    Bacillus atrophaeus cell line 9372: traditional spore strip 106 (24 month shelf life) / sterilisation process: dry heat
  • Length:
    30 mm
  • Seq:
    04