Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Fluorophenyl)propionic acid 96%

3-(2-Fluorophenyl)propionic acid 96%

Glass bottle for solids, 1 g
Catalog # H34110.03
Irritant
3-(2-Fluorophenyl)propionic acid 96%
Catalog # H34110.03
CAS Number:  1643-26-1
3-(2-Fluorophenyl)propionic acid 96%
3-(2-Fluorophenyl)propionic acid 96%

Glass bottle for solids, 1 g

CAS Number:  1643-26-1
Price
42,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 96% CAS No.: 1643-26-1 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 75-79° MDL No.: MFCD01310820

  • CAS Number:1643-26-1
  • Formula:C₉H₉FO₂
  • MDL Number:MFCD01310820
  • Molecular Weight:168.17 g/mol
  • Melting Pt:75…79 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.