3-(2-Fluorophenyl)propionic acid 96%
Glass bottle for solids, 250 mg
Supplier: Thermo Fisher Scientific
3-(2-Fluorophenyl)propionic acid 96%
Catalog # H34110.MD
Supplier: Thermo Fisher Scientific
CAS Number: 1643-26-1
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 1643-26-1 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 75-79° MDL No.: MFCD01310820
- CAS Number:1643-26-1
- Formula:C₉H₉FO₂
- MDL Number:MFCD01310820
- Molecular Weight:168.17 g/mol
- Melting Pt:75…79 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



