ContactUsLinkComponent
62 COMPOSES DANS HNO3 5%,HF 0,2% Se 1250mg/l Ca 1000mg/l P 1000mg/l Si 1000mg/l Be 500mg/l Te 500mg/l Ti 500mg/l As 250mg/l B 200mg/l Cu 150mg/l Ge 150mg/l Ni 150mg/l Zn 150mg/l Cd 100mg/l Li 100mg/l Mo 100mg/l Pd 100 1 * 100 mL
Catalog # CMPL1337.40.5N02F
62 COMPOSES DANS HNO3 5%,HF 0,2% Se 1250mg/l Ca 1000mg/l P 1000mg/l Si 1000mg/l Be 500mg/l Te 500mg/l Ti 500mg/l As 250mg/l B 200mg/l Cu 150mg/l Ge 150mg/l Ni 150mg/l Zn 150mg/l Cd 100mg/l Li 100mg/l Mo 100mg/l Pd 100 1 * 100 mL
Catalog # CMPL1337.40.5N02F