Order Entry
Germany
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Blue, 24 (3×8), 30 mm, 110×285,5×86,7 mm
  212-0425
VWR®, Test Tubes Racks
  212-0425
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Blue, 24 (3×8), 30 mm, 110×285,5×86,7 mm

 
32,60 €
   
1 * 1 ST
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    30 mm
  • No. of Holes:
    24 (3×8)
  • Colour:
    Blue
  • Seq:
    13
  • W×D×H:
    110×285,5×86,7 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.