Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR®, Test Tubes Racks

VWR®, Test Tubes Racks

Yellow, 72 (6×12), 16 mm, 127,3×250,6×73 mm
Catalog # 212-0412
Supplier:   VWR Collection
VWR®, Test Tubes Racks
Catalog # 212-0412
Supplier:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Yellow, 72 (6×12), 16 mm, 127,3×250,6×73 mm

Price
20,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    16 mm
  • No. of Holes:
    72 (6×12)
  • Colour:
    Yellow
  • Seq:
    60
  • W×D×H:
    127,3×250,6×73 mm