Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Adapter cable E-F, for connecting electrodes with type E plug (=DIN plug) to instruments with type F socket (mini-socket)
Catalog # METR6.2104.000
Supplier:   METROHM
Electrode Cables
Catalog # METR6.2104.000
Supplier:  METROHM
Electrode Cables
Adapter cable E-F, for connecting electrodes with type E plug (=DIN plug) to instruments with type F socket (mini-socket)
Price
362,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Adapter cable E-F, for connecting electrodes with type E plug (=DIN plug) to instruments with type F socket (mini-socket)
  • Seq:
    14