Order Entry
Germany
Orders LinkContactUsLinkComponent
COD standards

COD standards

COD standard solution, 50 mg/l, 500 mL
Catalog # REGECOD50
Supplier:   REAGECON
COD standards
Catalog # REGECOD50
Supplier:  REAGECON
CAS Number:  
COD standards

COD standard solution, 50 mg/l, 500 mL

CAS Number:  
Price
177,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Ideal for use as control standards to verify that correct analysis for COD has taken place.

Documents

Specifications

Specifications

  • Concentration:
    50 mg/l
  • Pk:
    500 mL
  • Description:
    COD standard solution

Specifications