Trypsin neutralizing solution
Trypsin neutralising solution (TNS), 30 mL
Catalog # C-41100
Marketsource
Supplier: PROMOCELL
Jump to:
Product Details & Documents
Trypsin is the most widely employed enzyme used for cell detachment. This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0,05% trypsin inhibitor in 0,1% BSA.
Caution: For in vitro research use only and not for diagnostic or therapeutic procedures.
Specifications
Specifications
- Pk:30 mL
- Description:Trypsin neutralising solution (TNS)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



