Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Human Recombinant AP1S2 (from E. coli)

Catalog # ABCAAB108122-250
Supplier:   Abcam
Human Recombinant AP1S2 (from E. coli)
Catalog # ABCAAB108122-250
Supplier:  Abcam
CAS Number:  
Human Recombinant AP1S2 (from E. coli)
Human Recombinant AP1S2 (from E. coli)
CAS Number:  
Price
2 290,00 €
Log in for availability
1 * 250 µG
Product Details & Documents

Recombinant Human AP1S2 protein is a Human Full Length protein, in the 1 to 157 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    AP1S2
  • Protein synonyms:
    DC22|AP-1 complex subunit sigma-2|Adaptor protein complex AP-1 subunit sigma-1B|Adaptor-related protein complex 1 subunit sigma-1B|Clathrin assembly protein complex 1 sigma-1B small chain|Golgi adaptor HA1/AP1 adaptin sigma-1B subunit|Sigma 1B subunit of AP-1 clathrin|Sigma-adaptin 1B|Sigma1B-adaptin
  • Purity:
    >85%
  • Concentration:
    0,5 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB108122-250
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 8; Constituents: 40% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0308% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Conjugation:
    Unconjugated
  • Tested applications:
    MS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications