Order Entry
Germany
Orders LinkContactUsLinkComponent
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld

Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld

Handheld pH/ISE/conductivity/DO meter Orion Star A329, instrument alone
Catalog # 665-0281
Supplier:   Thermo Orion
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld
Catalog # 665-0281
Supplier:  Thermo Orion
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld

Handheld pH/ISE/conductivity/DO meter Orion Star A329, instrument alone

Price
2 030,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This rugged, waterproof portable meter is designed for a wide range of pH, ion concentration, conductivity, dissolved oxygen and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP67-rated housing. The meter offers three measuring channels allowing users to measure pH, conductivity and DO simultaneously or view each channel separately.

  • Up to five point pH calibrations with calibration editing option that allows individual points to be fixed without a full recalibration
  • Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections

Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two.

Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings. Transfer single, range or all data points to printer or computer.

Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate. Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures. 3-years replacement warranty.

Certifications: TUV 3-1, FCC Class A.

Delivery information: Supplied with four with 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8107UWMMD ROSS Ultra Triode gel filled epoxy-body pH/ATC electrode (3 m cable), 013010MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor (3 m cable), 087010MD RDO optical DO sensor (3 m cable), calibration sleeve and guard, Orion pH 4/7/10 buffer pouches (10 packs each), ROSS electrode storage solution (475 ml), Orion rinse solution pouches (10 packs), Orion 1413 µS conductivity standard pouches (10 packs), Orion Star A series portable meter armor and Orion Star A series hard-sided field case.

Specifications
  • Description:
    Handheld pH/ISE/conductivity/DO meter Orion Star A329, instrument alone
  • Connections:
    BNC, pin tip, 8- and 9-pin mini-DIN
  • Battery type:
    4 AA (included)
  • Compliance:
    IP 67
  • DO concentration resolution:
    0,1 / 0,01 mg/L
  • Memory:
    Non-volatile memory preserves data log, calibration log and meter settings
  • ISE accuracy:
    ±0,2 mV or ±0,05% of reading, whichever is greater
  • IP rating:
    IP 67
  • DO accuracy:
    Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/l (up to 8 mg/), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
  • Data logging capacity:
    5000
  • Languages:
    English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • mV resolution:
    0,1
  • Resistivity range:
    2 Ω/cm - 100 MΩ/cm
  • Conductivity accuracy:
    ±0,5% of reading ±1 digit
  • DO calibration:
    Water-saturated air, air saturated water, manual (Winkler) and zero point
  • Pressure resolution:
    1 mm Hg
  • Input:
    3 measuring channels
  • ISE calibration:
    Up to 5 points
  • Resistivity resolution:
    1 ohm or 0,1 meg-ohm, auto ranging
  • TDS conversion factor:
    Linear (0,02 - 9,99)
  • W×D×H:
    60×105×240 mm
  • Temperature range:
    –5,0...+105,0 °C (–5,0...+50,0 °C with DO)
  • ISE range:
    0,0001 - 19900
  • Salinity range:
    0,06 - 80,00 psu / 0,05 - 42,00 ppt
  • mV range:
    ±2000,0
  • Conductivity reference temperature:
    5/10/15/20/25 °C
  • DO barometric pressure range:
    Automatic or manual, 450 - 850 mmHg
  • Resistivity accuracy:
    0,5% of reading ±1 digit
  • pH range:
    –2,000...+20,000
  • Pressure range:
    400 - 850 mm Hg
  • DO concentration:
    0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • Temperature accuracy:
    ±0,1 °C
  • Weight:
    0,5 kg
  • Cat. No.:
    665-0281
  • ISE units:
    ppm, M, mg/l, %, ppb, none
  • Outputs:
    RS232, USB
  • TDS range:
    0,001 - 200,0 ppm
  • mV accuracy:
    ±0,2 mV or 0,05%
  • ISE resolution:
    0,0001 minimum, 1 to 3 significant digits (user selectable)
  • Battery life:
    800 hours
  • Conductivity cell constant:
    0,001 - 199,9 cm⁻¹
  • Conductivity resolution:
    0,001 μS minimum, auto ranging, up to 4 significant digits
  • Power supply:
    AC adapter or 4 AA batteries
  • Salinity resolution:
    0,01 psu or 0,01 ppt minimum, auto ranging
  • TDS accuracy:
    0,5% of reading ±1 digit
  • Conductivity calibration:
    Up to 5 points
  • Temperature calibration:
    1 point temperature offset calibration
  • No. of channels:
    3
  • Calibration:
    pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • Temperature compensation:
    Automatic or manual
  • pH calibration:
    Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Temperature resolution:
    0,1 °C
  • TDS resolution:
    0,001 ppm minimum, auto ranging, up to 4 significant digits
  • Salinity accuracy:
    0,5% of reading ±1 digit
  • pH accuracy:
    ±0,002
  • pH resolution:
    0,1 / 0,01 / 0,001
  • Data logging - log function:
    Manual / Automatic with AUTO-READ™
  • Alarm:
    High/low limit alarm, calibration due alarm
  • Model:
    A329
  • Conductivity range:
    0,001 µS/cm - 3000 mS/cm
  • Calibration points:
    1 point relative mV (RmV) mode, 1 point ORP mode
  • DO saturation:
    0 - 600 % (polarographic); 0 - 500 % (RDO)
  • Display:
    Graphic LCD with backlight
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    662-1140
    Thermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters
    Log in for availability
    1 * 1 SET - 76,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Orion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve
    Orion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve
    ORIOSTARA-ESPH
    Log in for availability
    1 * 1 ST - 29,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve
    Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve
    ORIOSTARA-CS
    Log in for availability
    1 * 1 ST - 144,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    662-1547
    Log in for availability
    1 * 1 ST - 72,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Orion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve
    Orion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve
    ORIOSTARA-ESCD
    Log in for availability
    1 * 1 ST - 29,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Protective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holder
    Protective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holder
    ORIOSTARA-AR
    Log in for availability
    1 * 1 ST - 102,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the field
    Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the field
    ORIO1210005
    Log in for availability
    1 * 1 ST - 161,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • RS232 cable, for connecting a meter to a PC or printer
    RS232 cable, for connecting a meter to a PC or printer
    ORIO1010053
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star meters
    Log in for availability
    1 * 1 ST - 45,40 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Cable adapter, for connecting the RS232 port of a meter to a USB port
    Cable adapter, for connecting the RS232 port of a meter to a USB port
    ORIO1010005
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus meters
    Log in for availability
    1 * 1 ST - 131,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.