Order Entry
Germany
Orders LinkContactUsLinkComponent
Scoops

Scoops

310 mm, 110 mm, Red
Catalog # SCRW4280319
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # SCRW4280319
Supplier:  SCHROEDER WILESCO
Scoops
Scoops

310 mm, 110 mm, Red

Price
6,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These coloured scoops made from PP are suitable for food contact.

Specifications
  • Width:
    110 mm
  • Colour:
    Red
  • Length:
    310 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.