ContactUsLinkComponent
Recombinant protein, emilin2( np_114437.2, 121 a.a. - 230 a.a.) partial recombinant protein with . 1 * 10 µG
Catalog # ABNOH00084034-Q01
Supplier:  Abnova
Recombinant protein, emilin2( np_114437.2, 121 a.a. - 230 a.a.) partial recombinant protein with . 1 * 10 µG
Catalog # ABNOH00084034-Q01
Supplier:  Abnova
Related Information