0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, lrrn1( -, 1 a.a. - 716 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
ABNOH00057633-P01
:
Abnova
Recombinant protein, lrrn1( -, 1 a.a. - 716 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
ABNOH00057633-P01
:
Abnova