0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, myoz1( np_067068.1, 200 a.a. - 299 a.a.) partial recombinant protein with . 1 * 10 µG
ABNOH00058529-Q01
:
Abnova
Recombinant protein, myoz1( np_067068.1, 200 a.a. - 299 a.a.) partial recombinant protein with . 1 * 10 µG
ABNOH00058529-Q01
:
Abnova
sds_ABNOH00058529-Q01.pdf
TDS_ABNOH00058529-Q01.pdf