0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, hpse( np_006656.1, 72 a.a. - 171 a.a.) partial recombinant protein with . 1 * 10 µG
ABNOH00010855-Q01
:
Abnova
Recombinant protein, hpse( np_006656.1, 72 a.a. - 171 a.a.) partial recombinant protein with . 1 * 10 µG
ABNOH00010855-Q01
:
Abnova
sds_ABNOH00010855-Q01.pdf
TDS_ABNOH00010855-Q01.pdf