ContactUsLinkComponent
Recombinant protein, pskh1( aah62616, 1 a.a. - 100 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00005681-Q01
Supplier:  Abnova
Recombinant protein, pskh1( aah62616, 1 a.a. - 100 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00005681-Q01
Supplier:  Abnova