0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, dbi( np_065438, 1 a.a. - 104 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
ABNOH00001622-Q01
:
Abnova
Recombinant protein, dbi( np_065438, 1 a.a. - 104 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
ABNOH00001622-Q01
:
Abnova
sds_ABNOH00001622-Q01.pdf
TDS_ABNOH00001622-Q01.pdf