0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, hes1( np_005515.1, 201 a.a. - 280 a.a.) partial recombinant protein with . 1 * 10 µG
ABNOH00003280-Q03
:
Abnova
Recombinant protein, hes1( np_005515.1, 201 a.a. - 280 a.a.) partial recombinant protein with . 1 * 10 µG
ABNOH00003280-Q03
:
Abnova
sds_ABNOH00003280-Q03.pdf
TDS_ABNOH00003280-Q03.pdf