0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, nmi( aah21987.1, 1 a.a. - 307 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00009111-P01
:
Abnova
Recombinant protein, nmi( aah21987.1, 1 a.a. - 307 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00009111-P01
:
Abnova