0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Recombinant protein, pygb( np_002853.2, 1 a.a. - 843 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00005834-P01
:
Abnova
Recombinant protein, pygb( np_002853.2, 1 a.a. - 843 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00005834-P01
:
Abnova