Order Entry
Germany
Orders LinkContactUsLinkComponent
Glass beads

Glass beads

Glass beads, special filling, 5 mm
Catalog # HECH91140109005
Glass beads
Catalog # HECH91140109005
Glass beads
Glass beads

Glass beads, special filling, 5 mm

Price
Inquire For Price
Log in for availability
1 * 20 kg
Product Details & Documents

Solid glass.

For packing distillation columns.

Specifications
  • Ø:
    5 mm
  • Description:
    Glass beads, special filling
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.