ContactUsLinkComponent
DPD solution 1.1 g/l (N,N-Diethyl-1,4-phenylenediamine sulfate) for determination of chlorine according to EN ISO 7393:2000, DEV G4 Content: 0,5 l 1 * 500 mL
  KRAF20217.2700
DPD solution 1.1 g/l (N,N-Diethyl-1,4-phenylenediamine sulfate) for determination of chlorine according to EN ISO 7393:2000, DEV G4 Content: 0,5 l 1 * 500 mL
  KRAF20217.2700