V₂O₅ in Salpetersaure etwa 1 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF12940.0000
Supplier: Bernd Kraft
V₂O₅ in Salpetersaure etwa 1 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF12940.0000
Supplier: Bernd Kraft
Frequently Bought Together
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
Capric Alcohol, Reference Standard
Supplier: USP
Flurprimidol
Supplier: EHRENSTORFER
MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
Supplier: LGC Standards PROMOCHEM
Polyethylene Terephthalate, Reference Standards
Supplier: USP
Cobalt standard solution, 1000 mg/L Co in 5% nitric acid (from Co), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
ALPRAZOLAM RELATED COMPOUND A 1 * 50 mg
Supplier: Merck
3,6-Dichloropicolinic acid
Supplier: EHRENSTORFER
Chitosan, Reference Standard
Supplier: USP
VWR®, Sulphur, Certified Reference Materials, in synthetic diesel
Supplier: VWR Chemicals
Tetrabromobisphenol A
Supplier: EHRENSTORFER
Eicosapentaenoic Acid, Reference Standards
Supplier: USP