(NH₄)₂SO₄ in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF13741.0000
Supplier: Bernd Kraft
(NH₄)₂SO₄ in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF13741.0000
Supplier: Bernd Kraft
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Crystal violet solution Reag. Ph. Eur. 1022901
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
Manganese standard solution, for ICP-MS, Pure Plus grade
Supplier: PerkinElmer
Maduramicin ammonium salt certified 1 * 0,1 g
Supplier: EHRENSTORFER
in verdünnter Salzsäure/Salpetersäure gemäß EPA 200.7 1 * 0,5 L
Supplier: Bernd Kraft
Ethanolamine, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Buffer solutions
Supplier: REAGECON