ICP concentrate nickel 10.000 g Ni/l NiO in nitric acid 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF12939.0000
Supplier: Bernd Kraft
ICP concentrate nickel 10.000 g Ni/l NiO in nitric acid 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF12939.0000
Supplier: Bernd Kraft
Frequently Bought Together
1-Bromo-2-chloroethane
Supplier: EHRENSTORFER
MCPB D6 (ring D3, methyl D3) 100 ng/µl 1 * 1,1 mL
Supplier: EHRENSTORFER
Gadolinium standard solution, 1000 mg/L Gd in 5% nitric acid (de Gd?O?), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
5-Chloroethylthiamine Hydrochloride 1 * 50 mg
Supplier: EHRENSTORFER
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
Disulfoton-Oxon-Sulfoxide 1 * 25 mg
Supplier: EHRENSTORFER
BUFFER SOLUTION PH1.68 / 0.01, 25C 1 * 500 mL
Supplier: REAGECON
GINSENOSIDE RB2 1 * 10 mg
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Isopyrazam
Supplier: Merck
Pesticide reference standard, Endrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER