Cu in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF13124.0000
Supplier: Bernd Kraft
Cu in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF13124.0000
Supplier: Bernd Kraft
Frequently Bought Together
(1R)-(+)-α-Pinene, Supelco®
Supplier: Merck
2-Ethylcaproic acid
Supplier: EHRENSTORFER
1-Bromo-2-chloroethane
Supplier: EHRENSTORFER
ARMEPAVINE, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
Supelco® Organic Reference Standard, Protein Standard Set
Supplier: Merck
Mannose, Reference Standard
Supplier: USP
Amaranth certified 1 * 0,25 g
Supplier: EHRENSTORFER
pH buffer solutions, Specpure®
Supplier: Thermo Fisher Scientific
Neomycin sulphate
Supplier: EHRENSTORFER
Diisodecyl phthalate
Supplier: EHRENSTORFER
Boswellia Serrata Extract, Reference Standard
Supplier: USP
Pharmaceutical secondary standard, L-serine, (CRM), Supelco®
Supplier: Merck
Tioxazafen 1 * 10 mg
Supplier: EHRENSTORFER
Buffer solution, colour coded, ready to use
Supplier: Bernd Kraft
Taurine, Reference Standard
Supplier: USP