KNO₃ in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF10912.0000
Supplier: Bernd Kraft
KNO₃ in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF10912.0000
Supplier: Bernd Kraft
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW