ICP concentrate iron 10.000 g Fe/l Fe in nitric acid 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF10911.0000
Supplier: Bernd Kraft
ICP concentrate iron 10.000 g Fe/l Fe in nitric acid 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF10911.0000
Supplier: Bernd Kraft
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
VWR®, Osmolality standards
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
pH buffer solutions, colour coded
Supplier: Hach
Phosphorus standard solution, 1000 mg/L P in water (from KH₂PO₄) standard for ICP
Supplier: Bernd Kraft
pH buffer solutions, Specpure®
Supplier: Thermo Fisher Scientific