ICP concentrate cobalt 10.000 g Co/lCo in nitric acid approx. 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF17107.0000
Supplier: Bernd Kraft
ICP concentrate cobalt 10.000 g Co/lCo in nitric acid approx. 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF17107.0000
Supplier: Bernd Kraft
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
C TECHNOLOGIES VPE MEASUREMENT STANDARD 1 * 1 KIT
Supplier: MARKET SOURCE PART PROCESS
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Accessories for pH/°C meter, handheld
Supplier: Testo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Refractive index standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
STROPHANTHIDIN, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 20 mg
Supplier: Merck
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
Supplier: EHRENSTORFER
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
Supplier: VWR Chemicals
Alcohol calibration standards, Ethanol, (CRM), Supelco®
Supplier: Merck
Conductivity standards, at 25 °C, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Butyl acetate
Supplier: Merck