ICP concentrate calcium 10.000 g Ca/lCaCO3 in nitric acid 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Artikelnr. KRAF10910.0000
Supplier: Bernd Kraft
ICP concentrate calcium 10.000 g Ca/lCaCO3 in nitric acid 2 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Artikelnr. KRAF10910.0000
Supplier: Bernd Kraft
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Hydrochloric acid 32%, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Wash bottles, narrow neck, colour-coded, Nalgene®
Supplier: Thermo Fisher Scientific
(NH₄)₂SO₄ in Salpetersaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Supplier: Bernd Kraft