ContactUsLinkComponent
Silicic acid standard solution 100 mg Si/l SiCl4 in sodium hydroxide solution 0.2 mol/l according to DIN 38405-21:1990, DEV D21 1 * 500 mL
Catalog # KRAF21502.2000
Supplier:  Bernd Kraft
Silicic acid standard solution 100 mg Si/l SiCl4 in sodium hydroxide solution 0.2 mol/l according to DIN 38405-21:1990, DEV D21 1 * 500 mL
Catalog # KRAF21502.2000
Supplier:  Bernd Kraft
Frequently Bought Together