Nitric acid approx. 1 mol/l - approx. 1 M solution contains 0.2 - 0.5 % polyvinyl alcohol Content: 2,5 l 1 * 2.500 mL
Artikelnr. KRAF05912.4000
Supplier: Bernd Kraft
Nitric acid approx. 1 mol/l - approx. 1 M solution contains 0.2 - 0.5 % polyvinyl alcohol Content: 2,5 l 1 * 2.500 mL
Artikelnr. KRAF05912.4000
Supplier: Bernd Kraft
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Orthoboric acid 20 g/l in aqueous solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Disinfectant solutions, Bacillol AF
Supplier: BODE CHEMIE
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals