Nitric acid approx. 1 mol/l - approx. 1 M solution contains 0.2 - 0.5 % polyvinyl alcohol Content: 1 l 1 * 1 L
Catalog # KRAF05912.3000
Supplier: Bernd Kraft
Nitric acid approx. 1 mol/l - approx. 1 M solution contains 0.2 - 0.5 % polyvinyl alcohol Content: 1 l 1 * 1 L
Catalog # KRAF05912.3000
Supplier: Bernd Kraft
Frequently Bought Together
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Laboratory bottles, narrow neck, with thread, DURAN®
Supplier: DWK Life Sciences
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Disinfectant solutions, Bacillol AF
Supplier: BODE CHEMIE
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
TLC plates, aluminium oxide, Alox
Supplier: MACHEREY-NAGEL