Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Pyridine, extra pure

5 L
Catalog # KRAF07051.5500
Flammable
Irritant
Supplier:   Bernd Kraft
Pyridine, extra pure
Catalog # KRAF07051.5500
Supplier:  Bernd Kraft
CAS Number:  110-86-1
Pyridine, extra pure
Pyridine, extra pure

5 L

CAS Number:  110-86-1
Price
419,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pk:
    5 L

Specifications