Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyridine, extra pure

Pyridine, extra pure

Glass bottle, 250 mL
Catalog # KRAF07051.1700
Flammable
Irritant
Supplier:   Bernd Kraft
Pyridine, extra pure
Catalog # KRAF07051.1700
Supplier:  Bernd Kraft
CAS Number:  110-86-1
Pyridine, extra pure
Pyridine, extra pure

Glass bottle, 250 mL

CAS Number:  110-86-1
Price
46,40 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.