ContactUsLinkComponent
Buffer solution pH 10 54 g NH4Cl + 350 ml NH3-solution 25 % + 2 g Mg-K-EDTA + 5 g Na2S * 9-H2O/l in water for complexometric titrati on Content: 1 l 1 * 1 L
Catalog # KRAF03301.3000
Supplier:  Bernd Kraft
Buffer solution pH 10 54 g NH4Cl + 350 ml NH3-solution 25 % + 2 g Mg-K-EDTA + 5 g Na2S * 9-H2O/l in water for complexometric titrati on Content: 1 l 1 * 1 L
Catalog # KRAF03301.3000
Supplier:  Bernd Kraft