Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Plastic bottle, 1 L
Catalog # KRAF21440.3000
Supplier:   Bernd Kraft
Tween® 20 (Polysorbate)
Catalog # KRAF21440.3000
Supplier:  Bernd Kraft
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

Plastic bottle, 1 L

CAS Number:  9005-64-5
Price
111,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.