Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Plastic bottle, 2.500 mL
Catalog # KRAF21443.4000
Supplier:   Bernd Kraft
Tween® 80 (Polysorbate)
Catalog # KRAF21443.4000
Supplier:  Bernd Kraft
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Plastic bottle, 2.500 mL

CAS Number:  9005-65-6
Price
233,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:∼100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1.062 g/cm³ (20 °C)
  • Flash Pt:>149 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.