Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Plastic bottle, 1 L
Catalog # KRAF21443.3000
Supplier:   Bernd Kraft
Tween® 80 (Polysorbate)
Catalog # KRAF21443.3000
Supplier:  Bernd Kraft
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Plastic bottle, 1 L

CAS Number:  9005-65-6
Price
118,00 €
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:∼100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1.062 g/cm³ (20 °C)
  • Flash Pt:>149 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.