Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Plastic bottle, 500 mL
Catalog # KRAF21443.2000
Supplier:   Bernd Kraft
Tween® 80 (Polysorbate)
Catalog # KRAF21443.2000
Supplier:  Bernd Kraft
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Plastic bottle, 500 mL

CAS Number:  9005-65-6
Price
66,80 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:∼100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1.062 g/cm³ (20 °C)
  • Flash Pt:>149 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL

Specifications