54 g NH₄Cl + 350 ml NH₃-Losung 25 % + 1 g Zn-Na-EDTA/l in Wasser zur komplexometrischen Titration 1 * 1 ST
Catalog # KRAF03098.6000
Supplier: Bernd Kraft
54 g NH₄Cl + 350 ml NH₃-Losung 25 % + 1 g Zn-Na-EDTA/l in Wasser zur komplexometrischen Titration 1 * 1 ST
Catalog # KRAF03098.6000
Supplier: Bernd Kraft
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection