Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution pH 1,09

Buffer solution pH 1,09

Buffer solution, 1,09 (25 °C), 10 L
Catalog # KRAF03853.6000
Supplier:   Bernd Kraft
Buffer solution pH 1,09
Catalog # KRAF03853.6000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution pH 1,09
Buffer solution pH 1,09

Buffer solution, 1,09 (25 °C), 10 L

CAS Number:  
Price
294,00 €
Log in for availability
1 * 10 L
Product Details & Documents
  • Stabilized

Zertifizierung: According to DIN 19267.

Specifications

Specifications

  • Pk:
    10 L
  • Description:
    Buffer solution
  • pH value:
    1,09 (25 °C)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.