Buffer solution pH 1,09
Buffer solution, 1,09 (25 °C), 500 mL
Catalog # KRAF03853.2000
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
- Stabilized
Zertifizierung: According to DIN 19267.
Specifications
Specifications
- Pk:500 mL
- Description:Buffer solution
- pH value:1,09 (25 °C)



