ContactUsLinkComponent
IC multi-ion standard solution 2 anions in sodium hydroxide 0.005 mol/l for ion chromatography1 Liter contains 1000 mg of each: form iate, acetate Content: 0,5 l 1 * 500 mL
Catalog # KRAF04807.2000
Supplier:  Bernd Kraft
IC multi-ion standard solution 2 anions in sodium hydroxide 0.005 mol/l for ion chromatography1 Liter contains 1000 mg of each: form iate, acetate Content: 0,5 l 1 * 500 mL
Catalog # KRAF04807.2000
Supplier:  Bernd Kraft
Frequently Bought Together